MAT113927640 Matrix Scientific

trans-Chloro(1-naphthyl)bis(triphenylphosphine)-nickel

  • MDL number MFCD06800434
  • Cas Number : 54806-25-6
  • Molecular Weight : 745.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/016569.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/016569.html
Availability details for
Quantity
  • Availability
  • Add to Cart