MAT113927668 Matrix Scientific

4-(4-Chlorophenoxy)-3-fluoroaniline

  • MDL number MFCD08687429
  • Cas Number : 946664-06-8
  • Molecular Weight : 237.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022774.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022774.html
Availability details for
Quantity
  • Availability
  • Add to Cart