MAT113927673 Matrix Scientific

2-(4-Chlorophenoxy)-4-methylaniline

  • MDL number MFCD08687434
  • Cas Number : 919118-71-1
  • Molecular Weight : 233.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022232.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022232.html
Availability details for
Quantity
  • Availability
  • Add to Cart