MAT113927676 Matrix Scientific

4-(2-Chlorophenoxy)-3-methylaniline

  • MDL number MFCD08687437
  • Cas Number : 946785-01-9
  • Molecular Weight : 233.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022668.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022668.html
Availability details for
Quantity
  • Availability
  • Add to Cart