MAT113927687 Matrix Scientific

2-(2-Chlorophenoxy)-5-(trifluoromethyl)aniline

  • MDL number MFCD08687448
  • Cas Number : 946727-99-7
  • Molecular Weight : 287.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022047.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022047.html
Availability details for
Quantity
  • Availability
  • Add to Cart