MAT113927745 Matrix Scientific

4-(Cyclohexylmethoxy)-3-(trifluoromethyl)aniline

  • MDL number MFCD08687506
  • Cas Number : 946697-85-4
  • Molecular Weight : 273.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022484.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022484.html
Availability details for
Quantity
  • Availability
  • Add to Cart