MAT113927749 Matrix Scientific

4-(Cyclopentylmethoxy)aniline

  • MDL number MFCD08687509
  • Cas Number : 946729-84-6
  • Molecular Weight : 191.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022413.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022413.html
Availability details for
Quantity
  • Availability
  • Add to Cart