MAT113927751 Matrix Scientific

4-(Cyclopentylmethoxy)-3-fluoroaniline

  • MDL number MFCD08687511
  • Cas Number : 946663-82-7
  • Molecular Weight : 209.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022719.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022719.html
Availability details for
Quantity
  • Availability
  • Add to Cart