MAT113927755 Matrix Scientific

4-(Cyclopentylmethoxy)-3-methylaniline

  • MDL number MFCD08687515
  • Cas Number : 946741-59-9
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022639.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022639.html
Availability details for
Quantity
  • Availability
  • Add to Cart