MAT113927807 Matrix Scientific

3-(3,4-Difluorophenoxy)piperidine

  • MDL number MFCD08687570
  • Cas Number : 946714-62-1
  • Molecular Weight : 213.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023234.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023234.html
Availability details for
Quantity
  • Availability
  • Add to Cart