MAT113927814 Matrix Scientific

4-(2,4-Difluorophenoxy)-2-(trifluoromethyl)aniline

  • MDL number MFCD08687577
  • Cas Number : 946698-40-4
  • Molecular Weight : 289.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022636.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022636.html
Availability details for
Quantity
  • Availability
  • Add to Cart