MAT113927842 Matrix Scientific

(3',4'-Dimethyl[1,1'-biphenyl]-2-yl)methanamine

  • MDL number MFCD08687614
  • Cas Number : 946714-10-9
  • Molecular Weight : 211.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021780.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021780.html
Availability details for
Quantity
  • Availability
  • Add to Cart