MAT113927904 Matrix Scientific

4-(2-Ethoxyethoxy)-3-fluoroaniline

  • MDL number MFCD08687694
  • Cas Number : 946742-20-7
  • Molecular Weight : 199.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022725.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022725.html
Availability details for
Quantity
  • Availability
  • Add to Cart