MAT113927905 Matrix Scientific

2-(2-Ethoxyethoxy)-4-methylaniline

  • MDL number MFCD08687695
  • Cas Number : 946683-23-4
  • Molecular Weight : 195.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022182.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022182.html
Availability details for
Quantity
  • Availability
  • Add to Cart