MAT113927978 Matrix Scientific

4-(2-Ethylphenoxy)-3-fluoroaniline

  • MDL number MFCD08687767
  • Cas Number : 946664-00-2
  • Molecular Weight : 231.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022759.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022759.html
Availability details for
Quantity
  • Availability
  • Add to Cart