MAT113928098 Matrix Scientific

1-(2-Fluoro-4-nitrophenyl)homopiperazine

  • MDL number MFCD08692379
  • Cas Number : 392660-27-4
  • Molecular Weight : 239.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/024309.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/024309.html
Availability details for
Quantity
  • Availability
  • Add to Cart