MAT113928150 Matrix Scientific

4-(Cyclohexylmethoxy)-3-fluoroaniline

  • MDL number MFCD08687497
  • Cas Number : 937598-45-3
  • Molecular Weight : 223.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022720.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022720.html
Availability details for
Quantity
  • Availability
  • Add to Cart