MAT113928161 Matrix Scientific

N-Cyclohexyl-2-(methylamino)benzamide

  • MDL number MFCD00121552
  • Cas Number : 348612-65-7
  • Molecular Weight : 232.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122-123
Country of Origin USA
Product Link http://www.matrixscientific.com/023888.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023888.html
Availability details for
Quantity
  • Availability
  • Add to Cart