MAT113928178 Matrix Scientific

4-[(2-Chlorophenoxy)methyl]piperidine

  • MDL number MFCD06246403
  • Cas Number : 63608-31-1
  • Molecular Weight : 225.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023078.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023078.html
Availability details for
Quantity
  • Availability
  • Add to Cart