MAT113928229 Matrix Scientific

2-(2-Chlorophenoxy)propanoic acid

  • MDL number MFCD00002644
  • Cas Number : 25140-86-7
  • Molecular Weight : 200.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/024084.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/024084.html
Availability details for
Quantity
  • Availability
  • Add to Cart