MAT113928230 Matrix Scientific

2-(2-Ethoxyethoxy)aniline

  • MDL number MFCD08687692
  • Cas Number : 3062-48-4
  • Molecular Weight : 181.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021808.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021808.html
Availability details for
Quantity
  • Availability
  • Add to Cart