MAT113928234 Matrix Scientific

4-(2-Ethoxyethoxy)aniline hydrochloride

  • MDL number MFCD08559353
  • Cas Number : 65999-71-5
  • Molecular Weight : 217.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023847.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023847.html
Availability details for
Quantity
  • Availability
  • Add to Cart