MAT113928268 Matrix Scientific

4-Chloro-3-methylaniline

  • MDL number MFCD00066332
  • Cas Number : 7149-75-9
  • Molecular Weight : 141.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-84
Purity 98
Country of Origin USA
Boiling point 241
Product Link http://www.matrixscientific.com/021407.html

Safety Information

hcodes Irritant/Irritant-Harmful
MSDS http://www.matrixscientific.com/021407.html
Availability details for
Quantity
  • Availability
  • Add to Cart