MAT113928369 Matrix Scientific

N*1*,N*1*-Diethyl-1-p-tolyl-ethane-1,2-diamine

  • MDL number MFCD08667756
  • Cas Number : 919032-99-8
  • Molecular Weight : 206.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021560.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021560.html
Availability details for
Quantity
  • Availability
  • Add to Cart