MAT113928370 Matrix Scientific

4-(2-Ethylphenoxy)aniline

  • MDL number MFCD02663353
  • Cas Number : 383126-80-5
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022445.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022445.html
Availability details for
Quantity
  • Availability
  • Add to Cart