MAT113928380 Matrix Scientific

4,6-Difluoroindole

  • MDL number MFCD01075213
  • Cas Number : 199526-97-1
  • Molecular Weight : 153.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/023757.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023757.html
Availability details for
Quantity
  • Availability
  • Add to Cart