MAT113928479 Matrix Scientific

2,4-Dimethoxybenzenesulfonamide

  • MDL number MFCD06147001
  • Cas Number : 51770-71-9
  • Molecular Weight : 217.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166-167
Purity 96
Country of Origin USA
Product Link http://www.matrixscientific.com/021410.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021410.html
Availability details for
Quantity
  • Availability
  • Add to Cart