MAT113928495 Matrix Scientific

3-Fluoro-4'-methoxy[1,1'-biphenyl]-4-amine

  • MDL number MFCD06802177
  • Cas Number : 893734-49-1
  • Molecular Weight : 217.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021710.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021710.html
Availability details for
Quantity
  • Availability
  • Add to Cart