MAT113928539 Matrix Scientific

2,2-Dimethyl-chroman-4-ylamine

  • MDL number MFCD08687615
  • Cas Number : 220634-41-3
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 268�c.
Country of Origin USA
Product Link http://www.matrixscientific.com/021506.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021506.html
Availability details for
Quantity
  • Availability
  • Add to Cart