MAT113929181 Matrix Scientific

4-(2-Fluorophenoxy)-3-methylaniline

  • MDL number MFCD08687893
  • Cas Number : 946698-56-2
  • Molecular Weight : 217.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022667.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022667.html
Availability details for
Quantity
  • Availability
  • Add to Cart