MAT113929183 Matrix Scientific

4-(3-Fluorophenoxy)-3-methylaniline

  • MDL number MFCD08687895
  • Cas Number : 946785-13-3
  • Molecular Weight : 217.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022681.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022681.html
Availability details for
Quantity
  • Availability
  • Add to Cart