MAT113929193 Matrix Scientific

4-(2-Fluorophenoxy)-3-(trifluoromethyl)aniline

  • MDL number MFCD08687910
  • Cas Number : 74338-19-5
  • Molecular Weight : 271.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022507.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022507.html
Availability details for
Quantity
  • Availability
  • Add to Cart