MAT113929196 Matrix Scientific

4-(4-Fluorophenoxy)-2-(trifluoromethyl)aniline

  • MDL number MFCD08687913
  • Cas Number : 946784-69-6
  • Molecular Weight : 271.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022614.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022614.html
Availability details for
Quantity
  • Availability
  • Add to Cart