MAT113929300 Matrix Scientific

[(4-Methoxy-benzyl)-methyl-amino]-acetic acidhydrochloride

  • MDL number MFCD08741491
  • Cas Number : 857018-50-9
  • Molecular Weight : 245.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021528.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021528.html
Availability details for
Quantity
  • Availability
  • Add to Cart