MAT113929327 Matrix Scientific

1-Methoxy-3-methyl-2-butanaminehydrochloride

  • MDL number MFCD08741492
  • Cas Number : 1185294-54-5
  • Molecular Weight : 153.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/015559.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/015559.html
Availability details for
Quantity
  • Availability
  • Add to Cart