MAT113929332 Matrix Scientific

4-(2-Methoxy-4-methylphenoxy)-3-methylphenylamine

  • MDL number MFCD08688053
  • Cas Number : 946698-83-5
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022707.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022707.html
Availability details for
Quantity
  • Availability
  • Add to Cart