MAT113929380 Matrix Scientific

4-(3-Methoxypropoxy)-3-methylaniline

  • MDL number MFCD08688110
  • Cas Number : 946785-95-1
  • Molecular Weight : 195.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022664.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022664.html
Availability details for
Quantity
  • Availability
  • Add to Cart