MAT113929382 Matrix Scientific

4-(3-Methoxypropoxy)-2-(trifluoromethyl)aniline

  • MDL number MFCD08688112
  • Cas Number : 946697-97-8
  • Molecular Weight : 249.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022584.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022584.html
Availability details for
Quantity
  • Availability
  • Add to Cart