MAT113929428 Matrix Scientific

Methyl 4-[4-amino-3-(trifluoromethyl)phenoxy]-benzoate

  • MDL number MFCD08688156
  • Cas Number : 946741-43-1
  • Molecular Weight : 311.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022621.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022621.html
Availability details for
Quantity
  • Availability
  • Add to Cart