MAT113929536 Matrix Scientific

2-Methyl-4-(4-methylphenoxy)aniline

  • MDL number MFCD08688265
  • Cas Number : 946699-75-8
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54
Country of Origin USA
Product Link http://www.matrixscientific.com/022865.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022865.html
Availability details for
Quantity
  • Availability
  • Add to Cart