MAT113929636 Matrix Scientific

2-(2-Methyl-piperidine-1-sulfonyl)-ethylamineoxalate

  • MDL number MFCD08741495
  • Cas Number : 1185304-34-0
  • Molecular Weight : 296.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021512.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021512.html
Availability details for
Quantity
  • Availability
  • Add to Cart