MAT113929706 Matrix Scientific

2-Methyl-4-(tetrahydro-2-furanylmethoxy)aniline

  • MDL number MFCD08688397
  • Cas Number : 926259-32-7
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022818.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022818.html
Availability details for
Quantity
  • Availability
  • Add to Cart