MAT113929748 Matrix Scientific

Methyl 3-bromo-4-hydroxybenzoate

  • MDL number MFCD06203850
  • Cas Number : 29415-97-2
  • Molecular Weight : 231.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-110
Country of Origin USA
Product Link http://www.matrixscientific.com/024178.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/024178.html
Availability details for
Quantity
  • Availability
  • Add to Cart