MAT113929772 Matrix Scientific

4-[1-(2-Methoxy-ethyl)-2,5-dimethyl-1H-pyrrol-3-yl]-thiazol-2-ylamine

  • MDL number MFCD05511011
  • Cas Number : 917748-66-4
  • Molecular Weight : 251.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021667.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021667.html
Availability details for
Quantity
  • Availability
  • Add to Cart