MAT113929798 Matrix Scientific

Isobutyl 2-(3-oxo-2-piperazinyl)acetate

  • MDL number MFCD07600982
  • Cas Number : 1025496-29-0
  • Molecular Weight : 214.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023928.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023928.html
Availability details for
Quantity
  • Availability
  • Add to Cart