MAT113929808 Matrix Scientific

3-(2-Fluoro-phenoxy)-propionic acid

  • MDL number MFCD01569309
  • Cas Number : 2967-72-8
  • Molecular Weight : 184.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109-110
Country of Origin USA
Product Link http://www.matrixscientific.com/021478.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021478.html
Availability details for
Quantity
  • Availability
  • Add to Cart