MAT113929918 Matrix Scientific

3-Methoxyphenylhydrazine hydrochloride

  • MDL number MFCD00044416
  • Cas Number : 39232-91-2
  • Molecular Weight : 174.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142�c)
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/021395.html

Safety Information

hcodes Toxic/Store in Freezer
MSDS http://www.matrixscientific.com/021395.html
Availability details for
Quantity
  • Availability
  • Add to Cart