MAT113929948 Matrix Scientific

4-(3-Methylphenoxy)piperidinehydrochloride

  • MDL number MFCD09064908
  • Cas Number : 65367-96-6
  • Molecular Weight : 227.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/013718.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/013718.html
Availability details for
Quantity
  • Availability
  • Add to Cart