MAT113929951 Matrix Scientific

2-(4-Methylphenoxy)propanoic acid

  • MDL number MFCD02684121
  • Cas Number : 22504-83-2
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/024065.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/024065.html
Availability details for
Quantity
  • Availability
  • Add to Cart