MAT113929978 Matrix Scientific

4-Methoxy-2-piperidin-1-yl-thiazole-5-carbaldehyde

  • MDL number MFCD08688104
  • Cas Number : 919016-57-2
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74
Country of Origin USA
Product Link http://www.matrixscientific.com/021593.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021593.html
Availability details for
Quantity
  • Availability
  • Add to Cart